VWR®, Karl Fischer NewGen Solvent E-FI for volumetric two-component KF titration
Lieferant: VWR Chemicals
Medium for volumetric two-component Karl Fischer titration (ethanol-based), Free of imidazole (free of CMR substances)
- free of imidazole
- free of CMR substances
Specification:
Suitability (for KF-Titration): complying
Water content: 0.010 - 0.020% (v/v)