Glutaraldehyd 50% in Wasser, GPR RECTAPUR®
Lieferant: VWR Chemicals
|
Gefahr
|
Synonyme:
Glutaral, Pentandial, 1,5-Pentandial
88452.230EA
31.5
EUR
88452.230
Glutaraldehyd 50% in Wasser, GPR RECTAPUR®
Glutaraldehyd
Formel:
C₅H₈O₂ Molecular Weight: 100,12 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00007025 CAS-Nummer: 111-30-8 UN: 2922 ADR: 6.1(8),II |
Spezifikation Testergebnisse
Appearance | Clear colorless or yellow liquid |
Assay (GC) | 47,5 - 52% |
Color scale | max. 50 APHA |
pH | 3,2 - 4,2 |
Methanol | max. 0,3% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...