Iron nickel oxide, tech.

Lieferant: Thermo Fisher Scientific
Gefahr

88530.09 88530.18
88530.09EA 64.8 EUR
88530.09 88530.18
Iron nickel oxide, tech.
Iron nickel oxide

CAS No.: 12168-54-6 Molecular Formula: NiFe2O4 Formula Weight: 234.39 Physical Form: Powder Density: 5.368 Odour: Odourless MDL No.: MFCD00016254

Formel: NiFe₂O₄
Dichte: 5,368
Lagertemperatur: Raumtemperatur
MDL: MFCD00016254
CAS-Nummer: 12168-54-6
EINECS: 235-335-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR