Sulfolan, Supelco®
Lieferant: Merck
|
Achtung
|
Synonyme:
Thiacyclopentan-1,1-dioxid, Tetrahydrothiophen-1,1-dioxid, Tetramethylensulfon
M-Clarity™ Qualitätsstufe = MQ100
88965-5ML-F
88965-5ML-FEA
116
EUR
88965-5ML-F
Sulfolan, Supelco®
Sulfolan
Formel:
C₄H₈O₂S Molecular Weight: 120,17 g/mol |
MDL:
MFCD00005484 CAS-Nummer: 126-33-0 EINECS: 204-783-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...