Kieselgel, orange/grün, Perlen, GPR RECTAPUR® 2 - 5 mm, mit Indikator

Lieferant: VWR Chemicals

89054.292EA 30.7 EUR
89054.292 89054.360
Kieselgel, orange/grün, Perlen, GPR RECTAPUR® 2 - 5 mm, mit Indikator
Silica gel

Our silica gel orange/green is used where products need to be protected from humidity during storage and transportation. The colour changes from orange to dark green when saturated.


  • Mixture of various indicators and free of dangerous materials
  • Colour change from orange to dark green
  • Regeneration at 120 °C for 4 h

Formel: SiO₂
Siedepunkt: 2230 °C (1013 hPa)
Schmelzpunkt: > 1600 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00011232
CAS-Nummer: 7631-86-9

Bestellen

Spezifikation Testergebnisse

Absorption capacity (H₂O; 25 °C; 50% RH) ≥20,0%
Residual humidity (150 °C) ≤3,0%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR