Phytochemical standard, Eriodictyol, analytical standard, Supelco®
Lieferant: Merck
89061-10MG
89061-10MGEA 268 EUR
89061-10MG
Phytochemical standard, Eriodictyol, analytical standard, Supelco®
Standards Organische Standards
Eriodictyol is a naturally occurring flavonoid found in citrus fruits. It is isolated from the Chinese herb, Dracocephalum rupestre. It possess antioxidant activity.
Verpackung: Bottomless glass bottle. Contents are inside inserted fused cone.
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=85b73ca39eef6378e02a128aebe27bb600801a8d&vid=85b73ca39eef6378e02a128aebe27bb600801a8d&prid=web&clid=SAPDAM