Multi-element Ion chromatography cation standard solution, certified, Supelco®
Lieferant: Merck
89316-50ML-F
89316-50ML-FEA
305
EUR
89316-50ML-F
Multi-element Ion chromatography cation standard solution, certified, Supelco®
Standards
Kationischer Standards
Mehrelement-Standards
10,0 mg/kg ±0,2% Li+, Na+, K+, Mg²⁺, Ca²⁺ each cation, certified reference material traceable to SI.