Aluminum di(sec-butoxide)acetoacetic ester chelate (≥8,4% Al), tech.

Lieferant: Thermo Scientific
Achtung

89349.36
89349.36EA 45.9 EUR
89349.36
Aluminum di(sec-butoxide)acetoacetic ester chelate (≥8,4% Al), tech.
Aluminum di(sec-butoxide)acetoacetic ester chelate

CAS No.: 24772-51-8 Molecular Formula: C14H27AlO5 Formula Weight: 302.30 Flash Point: 76°(169°F) Physical Form: Liquid Density: 1.03 Sensitivity: Moisture Sensitive Odour: Alcohol-like MDL No.: MFCD00084762

Formel: C₁₄H₂₇AlO₅
Molecular Weight: 302,30 g/mol
Dichte: 1,03
Flammpunkt: 76 °C (169 °F)
Lagertemperatur: Raumtemperatur
MDL: MFCD00084762
CAS-Nummer: 24772-51-8
EINECS: 246-455-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR