Ammoniumsulfat ≥99,95% (Metall-Basis)

Lieferant: Thermo Fisher Scientific
Achtung

Synonyme: Diammoniumsulfat, di-Ammoniumsulfat

89363.22 89363.36 89363.A3
89363.22EA 40.7 EUR
89363.22 89363.36 89363.A3
Ammoniumsulfat ≥99,95% (Metall-Basis)
Ammoniumsulfat

Purity: 99.95% CAS No.: 7783-20-2 Molecular Formula: H8N2O4S Formula Weight: 132.14 Physical Form: Crystalline Density: 1.769 Appearance: White Melting Point: ca 280° dec. Merck Reference: 14,555 MDL No.: MFCD00003391

Formel: (NH₄)₂SO₄
Molecular Weight: 132.14 g/mol
Schmelzpunkt: ∼ 280 °C (Zers.)
Dichte: 1.769
Lagertemperatur: Raumtemperatur
MDL: MFCD00003391
CAS-Nummer: 7783-20-2
EINECS: 231-984-1
REACH: 01-2119455044-46
Merck Index: 14,555

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR