Kieselgel amorph, gedampft S.A. 350-420m2/g

Lieferant: Thermo Fisher Scientific
Achtung

Synonyme: Silica gel amorphous, Silicon dioxide amorphous, Silicium dioxide amorphous

89376.18 89376.30 89376.A1
89376.18EA 37 EUR
89376.18 89376.30 89376.A1
Kieselgel amorph, gedampft S.A. 350-420m2/g
Kieselgel amorph

CAS No.: 7631-86-9 Molecular Formula: SiO2 Formula Weight: 60.09 Physical Form: -325 Mesh Powder Density: 2.2 Refractive Index: 1.46 Odour: Odourless Melting Point: 1710° Boiling Point: 2230° Merck Reference: 14,8493 MDL No.: MFCD00011232

Formel: SiO₂
Molecular Weight: 60.08 g/mol
Siedepunkt: 2230 °C
Schmelzpunkt: 1710 °C
Dichte: 2.2
Lagertemperatur: Raumtemperatur
MDL: MFCD00011232
CAS-Nummer: 7631-86-9
EINECS: 231-545-4
Merck Index: 14,8493

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR