Bismut(III)chlorid, wasserfrei ≥99,999% (Metall-Basis)
Lieferant: Thermo Fisher Scientific
|
Gefahr
|
Purity: 99.999%
CAS No.: 7787-60-2
Molecular Formula: BiCl3
Formula Weight: 315.34
Physical Form: Crystalline
Density: 4.75
Appearance: White
Melting Point: 230-232°
Boiling Point: ca 430° subl.
Merck Reference: 14,1262
UN No.: 3260
MDL No.: MFCD00003461
Formel:
BiCl₃ Siedepunkt: ∼430 °C (Subl.) Schmelzpunkt: 230…232 °C Dichte: 4,75 Lagertemperatur: Raumtemperatur |
MDL:
MFCD00003461 CAS-Nummer: 7787-60-2 EINECS: 232-123-2 UN: 3260 ADR: 8,III Merck Index: 14,1262 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...