Ammonium-Stickstoff Standard für IC, TraceCERT®, Supelco®
Lieferant: Merck
89503-100ML
89503-100MLEA
56.1
EUR
89503-100ML
Ammonium-Stickstoff Standard für IC, TraceCERT®, Supelco®
Standards
Kationischer Standards
Ammonium-Standards
Standards: This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025.