Kupfer(II)-tetrafluoroborat 45% in wässriger Lösung

Lieferant: Thermo Scientific
Gefahr

Synonyme: Copper(II) borofluoride

89666.A1
89666.A1EA 93.6 EUR
89666.A1
Kupfer(II)-tetrafluoroborat 45% in wässriger Lösung
Copper(II) tetrafluoroborate

CAS No.: 38465-60-0 Molecular Formula: Cu(BF4)2 Formula Weight: 237.15 Physical Form: Liquid Appearance: Blue UN No.: UN3264 MDL No.: MFCD00016054

Formel: B₂CuF₈
Molecular Weight: 237,15 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00016054
CAS-Nummer: 38465-60-0
EINECS: 253-959-4
UN: 3264
ADR: 8,II

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR