Gold-Platin-Palladium ≥99,9% (Metall-Basis), Pulver APS <5 micron
Lieferant: Thermo Fisher Scientific
|
Achtung
|
89726.03
89726.03EA
293
EUR
89726.03
Gold-Platin-Palladium ≥99,9% (Metall-Basis), Pulver APS <5 micron
Gold platinum palladium
Purity: 99.9%
Molecular Formula: Au:Pt:Pd; 40:40:20 wt%
UN No.: UN3089
MDL No.: MFCD00192403
Formel:
Au:Pt:Pd;₄₀:₄₀:₂₀% Lagertemperatur: Raumtemperatur |
MDL:
MFCD00192403 UN: 3089 ADR: 4.1 III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...