O-Methylhydroxylamin Hydrochlorid 97,5-102,5% (durch argentometrische Titration), LiChropur™ zur GC-Derivatisierung, Supelco®

Lieferant: Merck
Gefahr

89803-1G 89803-5G
89803-1GEA 41.8 EUR
89803-1G 89803-5G
O-Methylhydroxylamin Hydrochlorid 97,5-102,5% (durch argentometrische Titration), LiChropur™ zur GC-Derivatisierung, Supelco®
O-Methylhydroxylamin Hydrochlorid
Formel: CH₆ClNO
MDL: MFCD00012951
CAS-Nummer: 593-56-6
UN: 3261
ADR: 8,II

Bestellen

Spezifikation Testergebnisse

Appearance (Color) White
Appearance (Form) Powder or Crystals
Argentometric Titration 97.5 - 102.5 %
Proton NMR Spectrum Conforms to Structure


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR