EDTA (Ethylendiamintetraessigsäure) 99,4-100,6%, Pulver, BAKER ANALYZED® ACS, J.T.Baker®
Lieferant: Avantor
|
Achtung
|
Synonyme:
Ethylendinitrilotetratetraessigsäure, Ethylendiamin-N,N,N',N'-tetraessigsäure, Kestranal® A, Edetinsäure, Idranal® II
8991-01EA
226.21
EUR
8991-01
EDTA (Ethylendiamintetraessigsäure) 99,4-100,6%, Pulver, BAKER ANALYZED® ACS, J.T.Baker®
EDTA (Ethylendiamintetraessigsäure)
The quality of many BAKER ANALYZED™ reagents meets or exceeds the requirements set forth by the American Chemical Society (ACS). These solvents are essential for any chemical stockroom.
Formel:
C₁₀H₁₆N₂O₈ Molecular Weight: 292,25 g/mol Siedepunkt: 614 °C (1013 hPa) Schmelzpunkt: 220 °C Dichte: 0,86 g/cm³ (20 °C) Flammpunkt: >100 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00003541 CAS-Nummer: 60-00-4 EINECS: 200-449-4 REACH: 01-2119486399-18 Merck Index: 13,03546 |
|
Spezifikation Testergebnisse
Meets ACS Reagent Chemical Requirements | 99.4 - 100.6 % |
For Laboratory,Research,or Manufacturing Use | 99.4 - 100.6 % |
ACS - Assay (C₁₀H₁₆N₂O₈) | 99.4 - 100.6 % |
ACS - Insoluble in Dilute NH₄OH | ≤0.005 % |
ACS - Residue after Ignition | ≤0.2 % |
ACS - Nitrilotriacetic Acid | ≤0.1 % |
ACS - Calcium (Ca) | ≤0.001 % |
ACS - Heavy Metals (as Pb) | ≤0.001 % |
ACS - Iron (Fe) | ≤0.005 % |
Trace Impurities - Magnesium (Mg) | ≤5 ppm |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...