EDTA Dinatriumsalz Dihydrat 99,0-101,0%, Kristalle, BAKER ANALYZED® ACS, J.T.Baker®
Lieferant: Avantor
The USP/NF Offical Compendia of Standards contains a section titled, "Reagents, Indicators, and Solutions" that lists the quality requirements of chemicals to be used in compendia testing. PharmaTest reagents are tested according to these requirements, relieving you of the need to do additional testing on your reagents. Products may be reagent or other grade, but are labeled as "Meets reagent specifications for testing USP/NF monographs." Test solutions are labeled as PharmaTest.
Formel:
C₁₀H₁₄N₂Na₂O₈·2H₂O Schmelzpunkt: 255 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00150037 CAS-Nummer: 6381-92-6 EINECS: 205-358-3 Merck Index: 13,03543 |
|
Spezifikation Testergebnisse
Meets ACS Reagent Chemical Requirements | |
For Laboratory,Research,or Manufacturing Use | 99.0 - 101.0 % |
Assay (C₁₀H₁₄N₂Na₂O₈·2H₂O) | 99.0 - 101.0 % |
Insoluble Matter | ≤0.005 % |
pH of 5% Solution at 25°C | 4.0 - 6.0 |
Nitrilotriacetic Acid (by polarography) | ≤0.1 % |
Heavy Metals (as Pb) | ≤0.005 % |
Iron (Fe) | ≤0.01 % |
Meets Reagent Specifications for testing USP/NF monographs |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...