Zinc molybdate ≥98%

Lieferant: Thermo Fisher Scientific

89937.06 89937.14
89937.06EA 67.4 EUR
89937.06 89937.14
Zinc molybdate ≥98%
Zinc molybdate

Purity: 98% CAS No.: 13767-32-3 Molecular Formula: ZnMoO4 Formula Weight: 225.32 Physical Form: Powder Melting Point: 700°C MDL No.: MFCD00049622

Formel: MoO₄Zn
Schmelzpunkt: >700 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00049622
CAS-Nummer: 13767-32-3
EINECS: 237-377-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR