Nickel molybdenum oxide ≥98%

Lieferant: Thermo Fisher Scientific
Gefahr

89938.14 89938.22
89938.14EA 54.9 EUR
89938.14 89938.22
Nickel molybdenum oxide ≥98%
Nickel molybdenum oxide

Purity: 98% CAS No.: 14177-55-0 Molecular Formula: NiMoO4 Formula Weight: 218.65 Physical Form: Powder Odour: Odourless MDL No.: MFCD00016255

Formel: NiMoO₄
Lagertemperatur: Raumtemperatur
MDL: MFCD00016255
CAS-Nummer: 14177-55-0
EINECS: 238-034-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR