di-Bortrioxid ≥99,98% (Metall-Basis)
Lieferant: Thermo Fisher Scientific
|
Gefahr
|
Purity: 99.98%
CAS No.: 1303-86-2
Molecular Formula: B2O3
Formula Weight: 69.62
Physical Form: Powder
Density: 2.46
Appearance: White
Odour: Odourless
Melting Point: ca 450°
Boiling Point: 1860°
Merck Reference: 14,1337
MDL No.: MFCD00011315
Formel:
B₂O₃ Siedepunkt: 1860 °C Schmelzpunkt: ∼450 °C Dichte: 2,46 Lagertemperatur: Raumtemperatur |
MDL:
MFCD00011315 CAS-Nummer: 1303-86-2 EINECS: 215-125-8 REACH: 01-2119486655-24 Merck Index: 14,1337 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...