Affinity chromatography media, Blue Sepharose™ High Performance
Lieferant: Cytiva
90-1001-23
90-1001-23EA
1695
EUR
90-1001-23
Affinity chromatography media, Blue Sepharose™ High Performance
Protein-Aufbereitungs
Protein Purification Resins
This media is suitable for affinity chromatography.
Chemical stability: Commonly used buffers, 70% ethanol, 8 M urea and 6 M guanidine hydrochloride.
Storage solution: 20% ethanol