Affinity chromatography media, Streptavidin Sepharose™ High Performance
Lieferant: Cytiva
90-1004-84
90-1004-84EA 2919 EUR
90-1004-84
Affinity chromatography media, Streptavidin Sepharose™ High Performance
Protein-Aufbereitungs Protein Purification Resins
Streptavidin Sepharose™ High Performance is an affinity chromatography resin designed for fast and reliable, high resolution purification of biotinylated biomolecules.
  • Extremely useful for exploiting either the strong interactions of biotin and streptavidin or the somewhat weaker interactions of 2-iminobiotin and streptavidin
  • 34 µm bead size for improved resolution
Purified Streptavidin isolated from Streptomyces avidinii is immobilised on Sepharose™ High Performance. The base matrix is a rigid, highly cross-linked beaded agarose with high chemical stability. The immobilised streptavidin binds biotin and biotinylated substances through affinity chromatography.