5(6)-Carboxyrhodamine 110 hydrochloride ≥90% (durch HPLC) Amin-reaktiver fluoreszierender Farbstoff

Lieferant: Biotium

Synonyme: 5(6)-CR 110 HCl

90000
90000.EA 281 EUR
90000.
5(6)-Carboxyrhodamine 110 hydrochloride ≥90% (durch HPLC) Amin-reaktiver fluoreszierender Farbstoff
5(6)-Carboxyrhodamine 110 hydrochloride

For some applications, 5-(and-6)-carboxyrhodamine (5(6)-CR 110) is a superior alternative to the widely used green fluorescent dye 5-(and-6)-carboxyfluorescein. The dye is insensitive to pH between 4 and 9 and is much more photostable than fluorescein.


  • λEx/λEm (MeOH) = 502/524 nm
  • Orange red solid soluble in H2O (pH>6), DMF or DMSO
  • Store at 4°C and protect from light

https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=814480909925cbe3a7a609b7c0d463b2ed0235c6&vid=1a89546cf02a05a8c4dbd35518cb21ac0c118a2e&prid=web&clid=SAPDAM

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR