5(6)-Carboxyrhodamine 110 succinimidyl ester ≥85% (durch HPLC) Amin-reaktiver fluoreszierender Farbstoff

Lieferant: Biotium

Synonyme: 5(6)-CR 110 SE

90001
90001.EA 425 EUR
90001.
5(6)-Carboxyrhodamine 110 succinimidyl ester ≥85% (durch HPLC) Amin-reaktiver fluoreszierender Farbstoff
5(6)-Carboxyrhodamine 110 succinimidyl ester

5-(and-6)-Carboxyrhodamine 110, SE (5-(and-6)-Carboxyrhodamine 110, succinimidyl ester, mixed isomers), also known as Carboxyrhodamine 110-NHS, is a superior green fluorescent dye that readily reacts with primary or secondary amines. For many applications, the dye is preferred over 5-(and-6)-carboxyfluorescein succinimidyl ester (5(6)-FAM) or FITC because of its exceptional photostability and fluorescence insensitivity to pH (4-9).


  • λEx/λ Em(MeOH) = 502/524 nm
  • Extinction Coefficient = 78,000
  • Orange red solid soluble in DMF or DMSO
  • Store at -20°C and protect from light

https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=cbc02507b59658001c24150cc16fd59e49a8dbcb&vid=4de7955b3241e27a51611ee7f125701ce77744da&prid=web&clid=SAPDAM

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR