5(6)-Carboxyrhodamine 110-X hydrochloride ≥95% (durch TLC) Amin-reaktiver fluoreszierender Farbstoff

Lieferant: Biotium

Synonyme: 5(6)-CR110-X HCl

90002
90002.EA 308 EUR
90002.
5(6)-Carboxyrhodamine 110-X hydrochloride ≥95% (durch TLC) Amin-reaktiver fluoreszierender Farbstoff
5(6)-Carboxyrhodamine 110-X hydrochloride

5-(and-6)-Carboxyrhodamine 110-X (full name: 5-(and-6)-Carboxyrhodamine 110-X, hydrochloride) is similar to 5-(and-6)-carboxyrhodamine 110 (#90000). The X linker molecule is useful to reduce the interaction between the dye and the molecule to be labeled.


  • λEx/λEm (MeOH) = 502/524 nm
  • Orange red solid soluble in water, DMF or DMSO
  • Store at 4°C and protect from light

https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=442c59edd1acf04ae3a334cd3e1e828a9cf5b3c6&vid=2f787626945d7f1e6a3b821622bd5296906771b3&prid=web&clid=SAPDAM

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR