5(6)-Carboxyrhodamine 110-X succinimudyl ester ≥90% (durch TLC) Amin-reaktiver fluoreszierender Farbstoff
Lieferant: Biotium
Synonyme:
5(6)-CR110-X SE
90003
90003.EA
470
EUR
90003.
5(6)-Carboxyrhodamine 110-X succinimudyl ester ≥90% (durch TLC) Amin-reaktiver fluoreszierender Farbstoff
5(6)-Carboxyrhodamine 110-X succinimudyl ester
5-(and-6)-Carboxyrhodamine 110-X, succinimidyl ester (5(6)-CR 110-X, SE) readily couples to a primary or secondary amine. The dye is a superior alternative to 5(6)-SFX because of its exceptional phototostability and fluorescence insensitivity to pH (4-9).
- λEx/λ Em (MeOH) = 502/524 nm
- Orange red solid soluble in DMSO or DMF
- Store at -20°C and protect from light
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...