5(6)-Carboxy-X-rhodamine triethylammonium salt ≥98% (durch TLC) Amin-reaktiver fluoreszierender Farbstoff

Lieferant: Biotium

Synonyme: 5(6)-ROX)

90004
90004.EA 169 EUR
90004.
5(6)-Carboxy-X-rhodamine triethylammonium salt ≥98% (durch TLC) Amin-reaktiver fluoreszierender Farbstoff
5(6)-Carboxy-X-rhodamine triethylammonium salt

5(6)-ROX (full name: 5-(and-6)-Carboxy-X-rhodamine, triethylammonium salt (mixed isomers)) is a long wavelength rhodamine dye.


  • λEx/λEm (MeOH) = 568/595 nm
  • Dark red solid soluble in DMSO, DMF or MeOH
  • Store at -20°C and protect from light

https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=0448e765e626b5d4fe667305efebab840bb62ee9&vid=0448e765e626b5d4fe667305efebab840bb62ee9&prid=web&clid=SAPDAM

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR