5-Carboxy-X-rhodamine triethylammonium salt einfaches Isomer ≥98% (durch TLC) Amin-reaktiver fluoreszierender Farbstoff
Lieferant: Biotium
Synonyme:
5-ROX
90006
90006.EA
242
EUR
90006.
5-Carboxy-X-rhodamine triethylammonium salt einfaches Isomer ≥98% (durch TLC) Amin-reaktiver fluoreszierender Farbstoff
5-Carboxy-X-rhodamine triethylammonium salt
5-ROX (full name: 5-Carboxy-X-rhodamine, triethylammonium salt, single isomer) is a single isomer, long wavelength rhodamine dye.
- λEx/λEm(MeOH) = 568/595 nm
- Dark red solid soluble in DMSO, DMF or MeOH
- Store at 4°C and protect from light
Spezifikation Testergebnisse
Molecular Weight | 635.6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...