6-Carboxy-X-rhodamine triethylammonium salt einfaches Isomer ≥95% (durch TLC) Amin-reaktiver fluoreszierender Farbstoff

Lieferant: Biotium

Synonyme: 6-ROX

90007
90007.EA 242 EUR
90007.
6-Carboxy-X-rhodamine triethylammonium salt einfaches Isomer ≥95% (durch TLC) Amin-reaktiver fluoreszierender Farbstoff
6-Carboxy-X-rhodamine triethylammonium salt

6-ROX (6-Carboxy-X-rhodamine) is a single isomer, long wavelength rhodamine dye.


  • λEx/λ Em(MeOH) = 568/595 nm
  • Dark red solid soluble in DMSO, DMF or MeOH
  • Store at 4°C and protect from light

https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=42df82180842e9da47f22c2a9c9bee38bfbf76e8&vid=ae119abc1811e32c105a2d15e4b4bc7800189641&prid=web&clid=SAPDAM

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR