5-Carboxytetramethylrhodamine (5-TAMRA) freie Säure Amin-reaktiver fluoreszierender Farbstoff

Lieferant: Biotium

90009
90009.EA 220 EUR
90009.
5-Carboxytetramethylrhodamine (5-TAMRA) freie Säure Amin-reaktiver fluoreszierender Farbstoff
5-Carboxytetramethylrhodamine (5-TAMRA)

Free acid form of 5-TAMRA (5-Carboxytetramethylrhodamine, single isomer).


  • λEx/λEm (MeOH) = 540/565 nm
  • Dark red solid soluble in DMSO, DMF, or MeOH
  • Store at 4°C and protect from light, especially in solution

Formel: C₂₅H₂₂N₂O₅
Lagertemperatur: Kühlschrank
MDL: MFCD00269769
CAS-Nummer: 91809-66-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=07d51c310d81ff6bdaa241fedb11103de876704b&vid=07d51c310d81ff6bdaa241fedb11103de876704b&prid=web&clid=SAPDAM

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR