6-Carboxyrhodamine 6G hydrochloride einfaches Isomer ≥95% (durch HPLC) Amin-reaktiver fluoreszierender Farbstoff

Lieferant: Biotium

Synonyme: 6-CR 6G

90011
90011.EA 186 EUR
90011.
6-Carboxyrhodamine 6G hydrochloride einfaches Isomer ≥95% (durch HPLC) Amin-reaktiver fluoreszierender Farbstoff
6-Carboxyrhodamine 6G hydrochloride

6-CR6G (full name: 6-Carboxyrhodamine 6G, hydrochloride, single isomer) is the free acid form of 6-carboxyrhodamine 6G single isomer.


  • λEx/λ Em (MeOH) = 520/546 nm
  • Orange red solid soluble in water (pH 7), DMF or DMSO
  • Store at 4°C and protect from light

https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=052537d0b3acccdd60c0765345aca31c634dc1dd&vid=04c55f4727e4481d84c706a0daaabc3627679074&prid=web&clid=SAPDAM

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR