5-Carboxyrhodamine 6G hydrochloride einfaches Isomer ≥95% (durch HPLC) Amin-reaktiver fluoreszierender Farbstoff

Lieferant: Biotium

Synonyme: 5-CR 6G

90012
90012.EA 186 EUR
90012.
5-Carboxyrhodamine 6G hydrochloride einfaches Isomer ≥95% (durch HPLC) Amin-reaktiver fluoreszierender Farbstoff
5-Carboxyrhodamine 6G hydrochloride

5-CR6G (full name: 5-Carboxyrhodamine 6G, hydrochloride, single isomer) is the free acid form of 5-carboxyrhodamine 6G single isomer.


  • λEx/λ Em(MeOH) = 520/546 nm
  • Orange red solid soluble in water (pH 7), DMF or DMSO
  • Store at 4°C and protect from light

https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=2e6429eb03d5ca9c405ae30ca028d8064491c564&vid=d81a22655862b06a7fa305d1529ce257ff58471d&prid=web&clid=SAPDAM

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR