6-ROX freie Säure ≥95% (durch HPLC) Amin-reaktiver fluoreszierender Farbstoff
Lieferant: Biotium
Synonyme:
6-Carboxy-X-rhodamine, Carboxy-X-rhodamine, 6-isomer
90013
90013.EA
236
EUR
90013.
6-ROX freie Säure ≥95% (durch HPLC) Amin-reaktiver fluoreszierender Farbstoff
6-ROX
6-Carboxy-X-rhodamine is a single isomer, long wavelength rhodamine dye.
- λEx/λEm(MeOH) = 568/595 nm
- Dark red solid soluble in DMSO, DMF or MeOH
- Store at -20°C and protect from light
Formel:
C₃₃H₃₀N₂O₅ Lagertemperatur: Gefrierschrank |
CAS-Nummer:
194785-18-7 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...