5-Carboxyrhodamine 110-X succinimidyl ester Amin-reaktiver fluoreszierender Farbstoff

Lieferant: Biotium

Synonyme: 5-CR110-X SE

90014
90014.EA 888 EUR
90014.
5-Carboxyrhodamine 110-X succinimidyl ester Amin-reaktiver fluoreszierender Farbstoff
5-Carboxyrhodamine 110-X succinimidyl ester

5-CR110-X, SE (full name: 5-Carboxyrhodamine 110-X, succinimidyl ester) is the amine-reactive form of 5-carboxyrhodamine 110 with a 6-carbon spacer.


  • λEx/λ Em (MeOH) = 502/524 nm
  • Orange red solid soluble in DMSO or DMF
  • Store at -20°C and protect from light

https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=249ec0536b8cadd8205bcba45754c1e82b12683f&vid=ac05c20522fcdfa17895f2bf4bbc31d7a8e0d043&prid=web&clid=SAPDAM

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR