Cyanine 555 succinimidyl ester Cyaninfarbstoff
Lieferant: Biotium
Biotium offers high quality amine-reactive succinimidyl ester (SE) dyes used to label proteins, oligonucleotides, and nucleic acids.
- Cyanine 555 is structurally identical to Cy®3 from GE Healthcare.
- λEx/λEm (water) = 555/565 nm
- Dark red solid soluble in DMF and DMSO
- Store at -20°C and protect from moisture and light
We offer Cyanine 555 (Cy®3), Cyanine 647 (Cy®5), Cyanine 680 (Cy®5.5), and Cyanine 750 (Cy®7) as high quality succinimidyl esters.
Also see our CF® Dye Succinimidyl Esters, available with more than 20 bright and photostable CF® Dyes.
Cy3, Cy5, Cy5.5, and Cy7 are registered trademarks of GE Healthcare.