Tetramethylrhodamine 6-isothiocyanate chloride ≥90% (durch TLC) Amin-reaktiver fluoreszierender Farbstoff
Lieferant: Biotium
Synonyme: 6-TRITC
90019
90019.EA 296 EUR
90019.
Tetramethylrhodamine 6-isothiocyanate chloride ≥90% (durch TLC) Amin-reaktiver fluoreszierender Farbstoff
Tetramethylrhodamine 6-isothiocyanate chloride (6-TRITC)
6-TRITC (Tetramethylrhodamine 6-isothiocyanate chloride) is spectrally similar to TAMRA (90022). The isothiocyanate reactive group reacts with primary or secondary amines to form a thiourea bond linkage.
  • Dark red solid soluble in DMF
  • Absorption/Emission 544nm/572nm (MeOH)
  • Extinction Coefficient >75,000
  • Store desiccated at -20 °C. Protect from light, especially when in solution.