6-Carboxyfluorescein-N-succinimidyl ester (6-FAM SE) ≥99% (durch TLC) Amin-reaktiver fluoreszierender Farbstoff
Lieferant: Biotium
Synonyme:
6-FAM SE, 6-Carboxyfluorescein NHS ester, 6-Carboxyfluorescein N-hydroxysuccinimide ester, 6-FAM-N-succinimidyl ester
90023
90023.EA
255
EUR
90023.
6-Carboxyfluorescein-N-succinimidyl ester (6-FAM SE) ≥99% (durch TLC) Amin-reaktiver fluoreszierender Farbstoff
6-Carboxyfluorescein-N-succinimidyl ester (6-FAM SE)
6-FAM, SE (full name: 6-Carboxyfluorescein, succinimidyl ester, single isomer) is the amine-reactive form of 6-carboxyfluorescein single isomer.
6-FAM SE is an amine-reactive green fluorescent dye widely used for labeling oligonucleotides or other biomolecules that contain a primary or secondary aliphatic amine. The coupling reaction is usually carried out at pH 8-9.5.
- λEx/λEm (pH 9) = 495/519 nm
- Yellow-orange solid soluble in DMF or DMSO
- Store at -20°C and protect from light
Formel:
C₂₅H₁₅NO₉ Molecular Weight: 473.39 g/mol Lagertemperatur: Gefrierschrank |
MDL:
MFCD00077323 CAS-Nummer: 92557-81-8 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...