5-Carboxyrhodamine 110 succinimidyl ester ≥90% (durch HPLC) Amin-reaktiver fluoreszierender Farbstoff

Lieferant: Biotium

Synonyme: 5-CR110 SE

90030
90030.EA 888 EUR
90030.
5-Carboxyrhodamine 110 succinimidyl ester ≥90% (durch HPLC) Amin-reaktiver fluoreszierender Farbstoff
5-Carboxyrhodamine 110 succinimidyl ester

Amine-reactive form of single isomer carboxyrhodamine 110, useful in DNA sequencing. For many single-dye-labeling applications, the dye is preferred over other green fluorescent dyes including fluorescein and Alexa 488 because of its exceptional photostability and fluorescence insensitivity to pH (4-9).


  • λEx/λ Em(MeOH) = 502/524 nm
  • Orange red solid soluble in DMF or DMSO
  • Store at -20°C and protect from light

https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f1cc84c1dc9989680826000431cf49dd3b22c1b6&vid=81ff56484de5a5f007c2ed9f670222dc6c58e25d&prid=web&clid=SAPDAM

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR