6-Carboxyrhodamine 110 succinimidyl ester ≥90% (durch HPLC) Amin-reaktiver fluoreszierender Farbstoff
Lieferant: Biotium
Synonyme:
6-CR110 SE
90031
90031.EA
888
EUR
90031.
6-Carboxyrhodamine 110 succinimidyl ester ≥90% (durch HPLC) Amin-reaktiver fluoreszierender Farbstoff
6-Carboxyrhodamine 110 succinimidyl ester
Amine-reactive form of single isomer carboxyrhodamine 110. For certain applications, the dye is preferred over the corresponding 6-carboxyfluorescein because of its exceptional photostability and fluorescence insensitivity to pH change (4-9).
- λEx/λ Em(MeOH) = 502/524 nm
- Orange red solid soluble in DMF or DMSO
- Store at -20°C and protect from light
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...