5-Carboxyrhodamine 6G succinimidyl ester ≥90% (durch HPLC) Amin-reaktiver fluoreszierender Farbstoff

Lieferant: Biotium

Synonyme: 5-CR 6G SE

90032
90032.EA 330 EUR
90032.
5-Carboxyrhodamine 6G succinimidyl ester ≥90% (durch HPLC) Amin-reaktiver fluoreszierender Farbstoff
5-Carboxyrhodamine 6G succinimidyl ester

Amine-reactive form of 5-carboxyrhodamine 6G single isomer.


  • λEx/λ Em(MeOH) = 520/546 nm
  • Orange red solid moderately soluble in DMSO
  • Store at -20°C and protect from light

https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8605cece7a81d0c2f6ee1869a385783c1f35fc0b&vid=7cf6a50312431ee2f7a8db2b23f33edece54055d&prid=web&clid=SAPDAM

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR