6-Carboxyrhodamine 6G succinimidyl ester (6-CR6G SE) ≥95% (durch TLC) Amin-reaktiver fluoreszierender Farbstoff

Lieferant: Biotium

Synonyme: Carboxyrhodamine 6G succinimidyl ester, 6-isomer, 6-CR6G, NHS ester, 6-Carboxyrhodamine 6G, succinimidyl ester

90033
90033.EA 330 EUR
90033.
6-Carboxyrhodamine 6G succinimidyl ester (6-CR6G SE) ≥95% (durch TLC) Amin-reaktiver fluoreszierender Farbstoff
6-Carboxyrhodamine 6G succinimidyl ester (6-CR6G SE)

Amine-reactive form of 6-carboxyrhodamine 6G single isomer.


  • λEx/λ Em(MeOH) = 520/546 nm
  • Orange red solid moderately soluble in DMSO
  • Store at -20°C and protect from light

Formel: C₃₁H₂₉N₃O₇
Molecular Weight: 555,59 g/mol
Lagertemperatur: Gefrierschrank
CAS-Nummer: 216699-37-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=193f79fe849e2a7456e810915e499999eb8a1df2&vid=193f79fe849e2a7456e810915e499999eb8a1df2&prid=web&clid=SAPDAM

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR