6-Carboxytetramethylrhodamine succinimidyl ester (6-TAMRA SE) ≥98% (durch TLC) Amin-reaktiver fluoreszierender Farbstoff

Lieferant: Biotium

Synonyme: 6-Carboxytetramethylrhodamine N-succinimidyl ester, 6-TAMRA, SE, 6-TAMRA N-succinimidyl ester

90035
90035.EA 325 EUR
90035.
6-Carboxytetramethylrhodamine succinimidyl ester (6-TAMRA SE) ≥98% (durch TLC) Amin-reaktiver fluoreszierender Farbstoff
6-Carboxytetramethylrhodamine succinimidyl ester (6-TAMRA SE)

Full name: 6-Carboxytetramethylrhodamine succinimidyl ester single isomer. Amine-reactive form ofcarboxytetramethylrhodamine (TAMRA) single isomer.


  • λEx/λEm (MeOH) = 540/565 nm
  • Dark red solid soluble in DMSO or DMF
  • Store at -20°C and protect from light

Formel: C₂₉H₂₅N₃O₇
Molecular Weight: 527.52 g/mol
Lagertemperatur: Gefrierschrank
CAS-Nummer: 150810-69-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=063801324835b30a328a4d6dbfd03295911af3d6&vid=063801324835b30a328a4d6dbfd03295911af3d6&prid=web&clid=SAPDAM

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR