5(6)-Carboxy-2′,7′-dichlorofluorescein diacetate N-succinimidyl ester (CDCFDA) ≥95% (durch TLC) fluoreszierender pH-Indikator
Lieferant: Biotium
Synonyme:
Carboxy-2′,7′- dichlorofluorescein diacetate, succinimidyl ester, 5 and 6 isomers, Carboxy-DCFDA-N-succinimidyl ester, CDCFDA
5-(and-6)-Carboxy-2′,7′-dichlorofluorescein diacetate, succinimidyl ester is a useful fluorescent tracer that can passively diffuse into cells and covalently label intracellular proteins, resulting in long-term cell labeling. The reagent itself is colorless and nonfluorescent but becomes brightly green fluorescent once it is hydrolyzed by intracellular esterases.
- λEx/λEm (of hydrolysis product at pH 7) = 504/529 nm
- pKa 4.8
- Cell permeability: Membrane permeant
- Colors: Green
- White solid soluble in DMSO or DMF
- Store at -20°C and protect from light
The fluorescence of the dye is relatively insensitive to pH change and the labeling has been shown to be stable to formaldehyde or gluteraldehyde fixation.
Formel:
C₂₉H₁₇Cl₂NO₁₁ Molecular Weight: 626,35 g/mol Lagertemperatur: Gefrierschrank |
CAS-Nummer:
147265-60-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...