6-(Fluorescein-5(6)-carboxamido) hexanoic acid succinimidyl ester [5(6)-SFX] Gemisch aus Isomeren ≥98% (durch TLC) Amin-reaktiver fluoreszierender Farbstoff

Lieferant: Biotium

90044
90044.EA 200 EUR
90044.
6-(Fluorescein-5(6)-carboxamido) hexanoic acid succinimidyl ester [5(6)-SFX] Gemisch aus Isomeren ≥98% (durch TLC) Amin-reaktiver fluoreszierender Farbstoff
6-(Fluorescein-5(6)-carboxamido) hexanoic acid succinimidyl ester [5(6)-SFX]

5(6)-SFX (full name: 6-(Fluorescein-5-(and-6)-carboxamido)hexanoic acid,succinimidyl ester) has a 6-atom aminohexanoyl spacer group that separates the fluorescein dye and the amine-reactive group.


  • λEx/λ Em(pH9) = 494/521nm
  • Yellow solid soluble in DMF, DMSO or pH>6.5 buffers
  • Store at -20°C and protect from light

Formel: C₃₁H₂₆N₂O₁₀
Lagertemperatur: Gefrierschrank
CAS-Nummer: 114116-31-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e252a7ed2d602c9b3209cd8f35f1d3b689ef774e&vid=e252a7ed2d602c9b3209cd8f35f1d3b689ef774e&prid=web&clid=SAPDAM

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR