4-Chlor-7-nitrobenzofurazan

Lieferant: Biotium

Synonyme: NBD chloride, 4-Chloro-7-Nitrobenz-2-Oxa-1,3-Diazole, NBD-Cl

90045
90045.EA 98.7 EUR
90045.
4-Chlor-7-nitrobenzofurazan
4-Chlor-7-nitrobenzofurazan
Formel: C₆H₂ClN₃O₃
Molecular Weight: 199,55 g/mol
MDL: MFCD00005808
CAS-Nummer: 10199-89-0
EINECS: 233-496-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR