Aceton ≥99,5% (durch GC, auf Wasserinhalt korrigiert), CMOS für Mikroelektronik, J.T.Baker®

Lieferant: Avantor
Gefahr

Synonyme: Propanon, DMK, 2-Propanon, Dimethylketon

9005-03EA 0 EUR
9005-03 9005-05
Aceton ≥99,5% (durch GC, auf Wasserinhalt korrigiert), CMOS für Mikroelektronik, J.T.Baker®
Aceton
Formel: CH₃COCH₃
Siedepunkt: 56,2 °C (1013 hPa)
Schmelzpunkt: –95,4 °C
Dichte: 0,792 g/cm³ (20 °C)
Flammpunkt: <–20 °C (geschlossener Tiegel)
Lagertemperatur: Raumtemperatur
MDL: MFCD00008765
CAS-Nummer: 67-64-1
EINECS: 200-662-2
UN: 1090
ADR: 3,II
REACH: 01-2119471330-49
Merck Index: 13,00067
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3de5d8e3bf7a94692d684c8f0ccdfb8500fde76d&vid=24fb2e0ec86974b1803780c7d5b23804a085df64&prid=web&clid=SAPDAM

Bestellen

Spezifikation Testergebnisse

For Microelectronic Use = 99.5 %
Assay ((CH₃)₂CO) (by GC, Corrected for Water) ≥99.5%
Colour (APHA) ≤10
Residue after Evaporation ≤5 ppm
Titrable Acid (µeq/g) ≤0.3
Titrable Base (µeq/g) ≤0.5
Water (H₂O) ≤0.5%
Solubility in H₂O Passes Test
Chloride (Cl) ≤0.2 ppm
Phosphate (PO₄) ≤0.05 ppm
Trace Impurities - Aluminum (Al) ≤50.0 ppb
Arsenic and Antimony (as As) ≤5.0 ppb
Trace Impurities - Barium (Ba) ≤20.0 ppb
Trace Impurities - Beryllium (Be) ≤10.0 ppb
Trace Impurities - Bismuth (Bi) ≤20.0 ppb
Trace Impurities - Boron (B) ≤10.0 ppb
Trace Impurities - Cadmium (Cd) ≤10.0 ppb
Trace Impurities - Calcium (Ca) ≤25.0 ppb
Trace Impurities - Chromium (Cr) ≤10.0 ppb
Trace Impurities - Cobalt (Co) ≤10.0 ppb
Trace Impurities - Copper (Cu) ≤10.0 ppb
Trace Impurities - Gallium (Ga) ≤10.0 ppb
Trace Impurities - Germanium (Ge) ≤10.0 ppb
Trace Impurities - Gold (Au) ≤20 ppb
Trace Impurities - Iron (Fe) ≤20.0 ppb
Trace Impurities - Lead (Pb) ≤10.0 ppb
Trace Impurities - Lithium (Li) ≤10.0 ppb
Trace Impurities - Magnesium (Mg) ≤20 ppb
Trace Impurities - Manganese (Mn) ≤10.0 ppb
Trace Impurities - Molybdenum (Mo) ≤10.0 ppb
Trace Impurities - Nickel (Ni) ≤10.0 ppb
Trace Impurities - Niobium (Nb) ≤50.0 ppb
Trace Impurities - Potassium (K) ≤10.0 ppb
Trace Impurities - Silicon (Si) ≤50 ppb
Trace Impurities - Silver (Ag) ≤10.0 ppb
Trace Impurities - Sodium (Na) ≤10.0 ppb
Trace Impurities - Strontium (Sr) ≤10.0 ppb
Trace Impurities - Tantalum (Ta) ≤50.0 ppb
Trace Impurities - Thallium (Tl) ≤10.0 ppb
Trace Impurities - Tin (Sn) ≤20.0 ppb
Trace Impurities - Titanium (Ti) ≤10.0 ppb
Trace Impurities - Vanadium (V) ≤10.0 ppb
Trace Impurities - Zinc (Zn) ≤20.0 ppb
Trace Impurities - Zirconium (Zr) ≤10.0 ppb
Particle Count - 0.5 µm and Greater (Rion KS42AF) ≤100 par/ml
Particle Count - 1.0 µm and Greater (Rion KS42AF) ≤8 par/ml


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR