Biotin ethylenediamine hydrobromide

Lieferant: Biotium

Synonyme: N-(2-Aminoethyl)biotinamide hydrobromide, Neurobiotin™, (3aS,4S,6aR)-N-(2-Aminoethyl)hexahydro- 2-oxo-1H-thieno[3,4-d]imidazole-4-pentanamide hydrobromide

90057
90057.EA 169 EUR
90057.
Biotin ethylenediamine hydrobromide
Biotin ethylenediamine hydrobromide

Biotin ethylenediamine, hydrobromide (N-(5-(6-((biotinoyl)amino)hexanoyl)amino)pentylmaleimide) is identical to Neurobiotin™, a useful anterograde and transneuronal tracer.


Neurobiotin is a trademark of Vector Laboratories.

Formel: C₁₂H₂₃BrN₄O₂S
Molecular Weight: 367.31 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD00467807
CAS-Nummer: 216299-38-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=1c72a727a6da20e6557d8c712bdf01341153a0e5&vid=9f6250f2fafe56c8317fa75d912ef19ad40feab2&prid=web&clid=SAPDAM

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR