Aceton ≥99,5% (durch GC, auf Wasserinhalt korrigiert), BAKER ANALYZED® ACS, J.T.Baker®

Lieferant: Avantor
Gefahr

Synonyme: Propanon, DMK, 2-Propanon, Dimethylketon

9006-01EA 46.51 EUR
9006-01 9006-R
Aceton ≥99,5% (durch GC, auf Wasserinhalt korrigiert), BAKER ANALYZED® ACS, J.T.Baker®
Aceton

The quality of many BAKER ANALYZED™ reagents meets or exceeds the requirements set forth by the American chemical society (ACS). These solvents are essential for any chemical stockroom.


  • Produced using proven distillation methods and rigid testing protocols
  • Statistical Process Control used in purification and packaging to ensure lot-to-lot consistency
  • Statistical Quality Control and suitability-for-use tests to ensure product performance

Formel: CH₃COCH₃
Siedepunkt: 56,2 °C (1013 hPa)
Schmelzpunkt: –95,4 °C
Dichte: 0,792 g/cm³ (20 °C)
Flammpunkt: <–20 °C (geschlossener Tiegel)
Lagertemperatur: Raumtemperatur
MDL: MFCD00008765
CAS-Nummer: 67-64-1
EINECS: 200-662-2
UN: 1090
ADR: 3,II
REACH: 01-2119471330-49
Merck Index: 13,00067
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3de5d8e3bf7a94692d684c8f0ccdfb8500fde76d&vid=24fb2e0ec86974b1803780c7d5b23804a085df64&prid=web&clid=SAPDAM

Bestellen

Spezifikation Testergebnisse

Meets ACS Reagent Chemical Requirements -
For Laboratory,Research,or Manufacturing Use = 99.5 %
Assay ((CH₃)₂CO) (by GC, corrected for water) ≥99.5 %
Color (APHA) ≤10
Residue after Evaporation ≤0.001 %
Solubility in H₂O Passes Test
Titrable Acid (meq/g) ≤0.0003
Titrable Base (meq/g) ≤0.0006
Aldehydes (as HCHO) ≤0.002 %
Isopropyl Alcohol (CH₃CHOHCH₃)(by GC) ≤0.05 %
Methanol (CH₃OH)(by GC) ≤0.05 %
Heavy Metals (as Pb) ≤1 ppm
Substances Reducing Permanganate Passes Test
Water (H₂O) ≤0.5 %
Copper (Cu) ≤0.1 ppm
Iron (Fe) ≤0.1 ppm
Nickel (Ni) ≤0.1 ppm
Date Received: __ __ /__ __ /__ __
Date Opened: __ __ /__ __ /__ __
Meets Reagent Specifications for testing USP/NF monographs


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR