Biocytin hydrazide

Lieferant: Biotium
Achtung

90060
90060.EA 240 EUR
90060.
Biocytin hydrazide
Biocytin hydrazide

Biocytin hydrazide is an aldehyde-fixable cellular tracer that can be introduced into cells by microinjection. Biocytin hydrazide can also be used to label glycoproteins or other carbohydrate molecules (See Reactive Dyes for Labeling Glycoproteins and Other Carbohydrate Molecules).


  • White solid soluble in DMSO
  • Store desiccated at 4°C

Formel: C₁₆H₃₀N₆O₃S
Lagertemperatur: Kühlschrank
CAS-Nummer: 102743-85-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR