Biotin ethylenediamine freie Base

Lieferant: Biotium

Synonyme: N-(2-Aminoethyl)biotinamide, (3aS,4S,6aR)-N-(2-Aminoethyl)hexahydro- 2-oxo-1H-thieno[3,4-d]imidazole-4-pentanamide

90071
90071.EA 146 EUR
90071.
Biotin ethylenediamine freie Base
Biotin ethylenediamine

Biotin ethylenediamine in free base form. White solid soluble in DMSO.

Formel: C₁₂H₂₂N₄O₂S
Molecular Weight: 286 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD00467807
CAS-Nummer: 216299-38-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=2be96bbbc5014834c5af8e2c292c4383aa7eebbe&vid=fbd00c3e229c46510e618e9dba3a2a624a022c18&prid=web&clid=SAPDAM

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR